Search
Search
| Gene Symbol: Ahsa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001115034.1 | NP_001108506.1 | 7 - 8 | 936 | 86 | |
| GenBank mRNA | BC158698.1 | - | 7 - 8 | 893 | 9027714 | 86 |
| FQ223544.1 | - | 7 - 8 | 920 | 86 | ||
| FQ232113.1 | - | 7 - 8 | 899 | 86 | ||
| FQ233084.1 | - | 7 - 8 | 898 | 86 | ||
Set Membership: |
|