Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01518137_m1
          See other ZFP410 GE Assays ›
          Gene Symbol:
          Zfp410
          Gene Name:
          zinc finger protein 410
          Gene Aliases:
          Znf410
          Chromosome Location:
          Chr.6: 108031734 - 108058836 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01518137_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp410
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001108042.2 NP_001101512.1 11 - 12 1591 92
          XM_006240338.1 XP_006240400.1 11 - 12 1612 92
          XM_006240339.1 XP_006240401.1 11 - 12 2523 92
          GenBank mRNA BC168180.1 - 11 - 12 1591 9100560 92

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0600001165.rn.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Process(es)

          nucleic acid binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P097799 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 544524 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 443421 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 928618 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 722655 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 313964 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 513048 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 154822 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 376460 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 229483 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 541878 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 363825 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 486011 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 826073 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 332410 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 190942 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 400491 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 790151 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 877220 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 159806 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 56345 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P097799 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P097799 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P097799 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P543930 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 21259 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10885909 - -
          Affymetrix Rat Gene 1.0 ST Array 10885913 - -
          Affymetrix Rat Gene 1.0 ST Array 10885914 - -
          Affymetrix Rat Gene 1.0 ST Array 10885912 - -
          Affymetrix Rat Gene 1.0 ST Array 10885911 - -
          Affymetrix Rat Gene 1.0 ST Array 10885915 - -
          Affymetrix Rat Gene 1.0 ST Array 10885908 - -
          Affymetrix Rat Gene 1.0 ST Array 10885907 - -
          Affymetrix Rat Gene 1.0 ST Array 10885910 - -
          Affymetrix Rat Gene 1.0 ST Array 10885916 - -
          Affymetrix Rat Gene 1.0 ST Array 10885917 - -
          Affymetrix Rat Gene 1.1 ST Array 10885906 - -
          Rat Genome 230 2.0 Array 1391093_at - -
          Rat Genome 230 2.0 Array 1392966_PM_at - -
          Rat Genome 230 2.0 Array 1391093_PM_at - -
          Rat Genome 230 2.0 Array 1392966_at - -
          Affymetrix HT Rat Focus Array 1392966_at - -
          Affymetrix HT Rat Focus Array 1391093_PM_at - -
          Affymetrix HT Rat Focus Array 1392966_PM_at - -
          Affymetrix HT Rat Focus Array 1391093_at - -
          Affymetrix Rat Clariom S Assay PSR0600008160.rn.1 TC0600001165.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600008157.rn.1 TC0600001165.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600008159.rn.1 TC0600001165.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0600008156.rn.1 TC0600001165.rn.1 RTA TC
          Rat Expression Set 230 1391093_at - -
          Rat Expression Set 230 1392966_PM_at - -
          Rat Expression Set 230 1391093_PM_at - -
          Rat Expression Set 230 1392966_at - -
          Affymetrix Rat Gene 1.0 ST Array 10885906 - -
          Rat Genome U34 Set rc_AA899749_at - -
          Rat Genome U34 Set rc_AI236169_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21994034 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1363652 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline