Search
Search
| Gene Symbol: Fbxo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053511.1 | NP_445963.1 | 3 - 4 | 600 | 80 | |
| XM_008764298.1 | XP_008762520.1 | 4 - 5 | 803 | 80 | ||
| GenBank mRNA | AF098301.1 | - | 3 - 4 | 600 | 80 | |
| FQ214021.1 | - | 3 - 4 | 593 | 80 | ||
Set Membership: |
|