Search
Search
| Gene Symbol: Wasf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013167.1 | NP_001013185.1 | 3 - 4 | 472 | 67 | |
| XM_008764138.1 | XP_008762360.1 | 4 - 5 | 653 | 67 | ||
| GenBank mRNA | BC089207.1 | - | 3 - 4 | 472 | 7135528 | 67 |
Set Membership: |
|