Search
Search
| Gene Symbol: Rcc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128189.2 | NP_001121661.1 | 8 - 9 | 979 | 118 | |
| XM_006239073.2 | XP_006239135.1 | 8 - 9 | 949 | 118 | ||
| XM_006239074.1 | XP_006239136.1 | 8 - 9 | 1273 | 118 | ||
| XM_006239075.2 | XP_006239137.1 | 8 - 9 | 1463 | 118 | ||
| XM_006239077.2 | XP_006239139.1 | 7 - 8 | 909 | 118 | ||
| XM_008764172.1 | XP_008762394.1 | 8 - 9 | 2617 | 118 | ||
| XM_008764173.1 | XP_008762395.1 | 9 - 10 | 1003 | 118 | ||
| GenBank mRNA | BC160834.1 | - | 8 - 9 | 979 | 7456254 | 118 |
Set Membership: |
|