Search
Search

LOC682102| Gene Symbol: LOC682102 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134732.1 | NP_001128204.1 | 6 - 7 | 1859 | 129 | |
| GenBank mRNA | BC168197.1 | - | 5 - 6 | 1166 | 7112960 | 129 |
Set Membership: |
|