Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01520745_m1
          See other ERRFI1 GE Assays ›
          Gene Symbol:
          Errfi1
          Gene Name:
          ERBB receptor feedback inhibitor 1
          Gene Aliases:
          RGD1307599, Ralt, Xxx, gene 33
          Chromosome Location:
          Chr.5: 167951561 - 167966011 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01520745_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Errfi1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001014071.1 NP_001014093.1 3 - 4 466 105
          XM_008764260.1 XP_008762482.1 2 - 3 3214 105
          XM_008764261.1 XP_008762483.1 3 - 4 550 105
          GenBank mRNA BC083845.1 - 3 - 4 466 7121187 105
          X07266.1 - 3 - 4 469 105

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500001969.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Errfi1

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of epidermal growth factor-activated receptor activity
          negative regulation of protein autophosphorylation
          negative regulation of interleukin-1 beta production
          cellular response to insulin stimulus
          negative regulation of collagen biosynthetic process
          cellular response to platelet-derived growth factor stimulus
          negative regulation of epidermal growth factor receptor signaling pathway
          negative regulation of tumor necrosis factor biosynthetic process
          skin morphogenesis
          regulation of keratinocyte differentiation
          lung alveolus development
          negative regulation of peptidyl-tyrosine phosphorylation
          lung vasculature development
          lung epithelium development
          regulation of type B pancreatic cell proliferation
          negative regulation of ERK1 and ERK2 cascade
          cellular response to epidermal growth factor stimulus
          cellular hyperosmotic response
          cellular response to dexamethasone stimulus
          negative regulation of cardiac muscle hypertrophy in response to stress
          protein binding
          SH3 domain binding
          kinase binding
          protein kinase binding
          small GTPase binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P14882 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 573726 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 577554 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 312822 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 514108 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 690209 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 918105 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 869944 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 501949 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 92134 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 93746 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P14882 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P14882 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P14882 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P14882 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700492544 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 50108 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 74218 - -
          Agilent Rat cDNA Microarray (G4105A) 700492544 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10874197 - -
          Affymetrix Rat Gene 1.0 ST Array 10874196 - -
          Affymetrix Rat Gene 1.0 ST Array 10874195 - -
          Affymetrix Rat Gene 1.0 ST Array 10874194 - -
          Affymetrix Rat Gene 1.1 ST Array 10874193 - -
          Rat Genome 230 2.0 Array 1373093_at - -
          Rat Genome 230 2.0 Array 1373093_PM_at - -
          Affymetrix HT Rat Focus Array 1373093_at - -
          Affymetrix HT Rat Focus Array 1373093_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0500014790.rn.1 TC0500001969.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500014789.rn.1 TC0500001969.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500014788.rn.1 TC0500001969.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500014786.rn.1 TC0500001969.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500014787.rn.1 TC0500001969.rn.1 RTA TC
          Rat Expression Set 230 1373093_at - -
          Rat Expression Set 230 1373093_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10874193 - -
          Rat Genome U34 Set M23572_at - -
          Rat Genome U34 Set X07266cds_s_at - -
          Rat Genome U34 Set rc_AI169756_s_at - -
          Rat Genome U34 Set rc_AI169231_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21399344 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1366212 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline