Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01520800_m1
          See other RSRP1 GE Assays ›
          Gene Symbol:
          Rsrp1
          Gene Name:
          arginine/serine-rich protein 1
          Gene Aliases:
          RGD1359529
          Chromosome Location:
          Chr.5: 153260930 - 153264669 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01520800_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rsrp1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001014193.2 NP_001014215.2 2 - 3 777 63
          GenBank mRNA BC085888.1 - 2 - 3 725 7192402 63
          CO405856.1 - 1 - 2 539 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500001668.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rsrp1

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P054193 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 283342 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 138546 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 601447 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 611977 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P054193 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P054193 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P054193 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P396370 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P261538 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 40758 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10872835 - -
          Affymetrix Rat Gene 1.0 ST Array 10872831 - -
          Affymetrix Rat Gene 1.0 ST Array 10872834 - -
          Affymetrix Rat Gene 1.0 ST Array 10872833 - -
          Affymetrix Rat Gene 1.0 ST Array 10872830 - -
          Affymetrix Rat Gene 1.0 ST Array 10872832 - -
          Affymetrix Rat Gene 1.1 ST Array 10872829 - -
          Rat Genome 230 2.0 Array 1398364_PM_at - -
          Rat Genome 230 2.0 Array 1390943_at - -
          Rat Genome 230 2.0 Array 1398364_at - -
          Rat Genome 230 2.0 Array 1390943_PM_at - -
          Affymetrix HT Rat Focus Array 1390943_at - -
          Affymetrix HT Rat Focus Array 1390943_PM_at - -
          Affymetrix HT Rat Focus Array 1398364_PM_at - -
          Affymetrix HT Rat Focus Array 1398364_at - -
          Affymetrix Rat Clariom S Assay PSR0500012045.rn.1 TC0500001668.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500012044.rn.1 TC0500001668.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500012043.rn.1 TC0500001668.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500012042.rn.1 TC0500001668.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500012046.rn.1 TC0500001668.rn.1 RTA TC
          Rat Expression Set 230 1398364_at - -
          Rat Expression Set 230 1398364_PM_at - -
          Rat Expression Set 230 1390943_at - -
          Rat Expression Set 230 1390943_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10872829 - -
          Rat Genome U34 Set rc_AI045196_at - -
          Rat Genome U34 Set rc_AI058906_at - -
          Rat Genome U34 Set rc_AA924413_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21337048 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1366485 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline