Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01520888_g1
          See other TARDBP GE Assays ›
          Gene Symbol:
          Tardbp
          Gene Name:
          TAR DNA binding protein
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 165432089 - 165442112 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01520888_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tardbp
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001011979.2 NP_001011979.1 5 - 6 815 94
          XM_006239382.2 XP_006239444.1 5 - 6 868 94
          XM_006239384.2 XP_006239446.1 5 - 6 845 94
          XM_006239385.2 XP_006239447.1 5 - 6 846 94
          XM_006239386.2 XP_006239448.1 5 - 6 850 94
          XM_008764234.1 XP_008762456.1 5 - 6 847 94
          XM_008764235.1 XP_008762457.1 5 - 6 845 94
          XM_008764236.1 XP_008762458.1 5 - 6 845 94
          GenBank mRNA BC083752.1 - 5 - 6 804 7192591 94

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500004055.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tardbp

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription from RNA polymerase II promoter
          RNA splicing
          positive regulation of insulin secretion
          response to endoplasmic reticulum stress
          negative regulation by host of viral transcription
          positive regulation of transcription from RNA polymerase II promoter
          3'-UTR-mediated mRNA stabilization
          nucleotide binding
          transcriptional activator activity, RNA polymerase II distal enhancer sequence-specific binding
          double-stranded DNA binding
          RNA binding
          mRNA 3'-UTR binding
          identical protein binding
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P040710 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P040715 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 756056 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 735255 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 406264 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 84838 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 425976 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 131123 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 840711 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 22941 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 649961 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 44475 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P040710 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P040715 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P040710 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P040715 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P040710 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P040715 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P335617 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 96182 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 17283 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10881640 - -
          Affymetrix Rat Gene 1.0 ST Array 10881639 - -
          Affymetrix Rat Gene 1.0 ST Array 10881634 - -
          Affymetrix Rat Gene 1.0 ST Array 10881641 - -
          Affymetrix Rat Gene 1.0 ST Array 10881633 - -
          Affymetrix Rat Gene 1.0 ST Array 10881642 - -
          Affymetrix Rat Gene 1.0 ST Array 10881635 - -
          Affymetrix Rat Gene 1.0 ST Array 10881637 - -
          Affymetrix Rat Gene 1.0 ST Array 10881631 - -
          Affymetrix Rat Gene 1.0 ST Array 10881638 - -
          Affymetrix Rat Gene 1.0 ST Array 10881632 - -
          Affymetrix Rat Gene 1.1 ST Array 10881630 - -
          Rat Genome 230 2.0 Array 1377608_a_at - -
          Rat Genome 230 2.0 Array 1371367_at - -
          Rat Genome 230 2.0 Array 1378259_at - -
          Rat Genome 230 2.0 Array 1371367_PM_at - -
          Rat Genome 230 2.0 Array 1377608_PM_a_at - -
          Rat Genome 230 2.0 Array 1378259_PM_at - -
          Affymetrix HT Rat Focus Array 1377608_a_at - -
          Affymetrix HT Rat Focus Array 1371367_PM_at - -
          Affymetrix HT Rat Focus Array 1378259_PM_at - -
          Affymetrix HT Rat Focus Array 1371367_at - -
          Affymetrix HT Rat Focus Array 1378259_at - -
          Affymetrix HT Rat Focus Array 1377608_PM_a_at - -
          Affymetrix Rat Clariom S Assay PSR0500030918.rn.1 TC0500004055.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500030919.rn.1 TC0500004055.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500030927.rn.1 TC0500004055.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500030922.rn.1 TC0500004055.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500030921.rn.1 TC0500004055.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500030932.rn.1 TC0500004055.rn.1 RTA TC
          Rat Expression Set 230 1371367_at - -
          Rat Expression Set 230 1371367_PM_at - -
          Rat Expression Set 230 1377608_a_at - -
          Rat Expression Set 230 1377608_PM_a_at - -
          Rat Expression Set 230 1378259_at - -
          Rat Expression Set 230 1378259_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10881630 - -
          Rat Genome U34 Set D84484_at - -
          Rat Genome U34 Set rc_AA944833_g_at - -
          Rat Genome U34 Set rc_AA944833_at - -
          Rat Genome U34 Set rc_AA999071_at - -
          Rat Genome U34 Set rc_AI175095_s_at - -
          Rat Genome U34 Set rc_AI232957_at - -
          Rat Genome U34 Set rc_AI236146_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376454 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline