Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01521728_g1
          See other PSRC1 GE Assays ›
          Gene Symbol:
          Psrc1
          Gene Name:
          proline/serine-rich coiled-coil 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 211176447 - 211181078 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01521728_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Psrc1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001044302.1 NP_001037767.1 7 - 8 1063 79
          XM_006233176.2 XP_006233238.1 7 - 8 1273 79
          XM_006233177.2 XP_006233239.1 7 - 8 1104 79
          XM_006233179.2 XP_006233241.1 7 - 8 1104 79
          GenBank mRNA BC105874.1 - 7 - 8 1063 7131142 79

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200001990.rn.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          microtubule bundle formation
          mitotic metaphase plate congression
          cellular process
          negative regulation of cell growth
          positive regulation of microtubule polymerization
          positive regulation of cyclin-dependent protein serine/threonine kinase activity
          positive regulation of transcription, DNA-templated
          cell division
          regulation of mitotic spindle organization
          microtubule binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P838583 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P061770 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 98280 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 705815 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 7983 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P061770 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P838583 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P061770 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P838583 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P838583 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P061770 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10818363 - -
          Affymetrix Rat Gene 1.0 ST Array 10818359 - -
          Affymetrix Rat Gene 1.0 ST Array 10818361 - -
          Affymetrix Rat Gene 1.0 ST Array 10818367 - -
          Affymetrix Rat Gene 1.0 ST Array 10818362 - -
          Affymetrix Rat Gene 1.0 ST Array 10818365 - -
          Affymetrix Rat Gene 1.0 ST Array 10818360 - -
          Affymetrix Rat Gene 1.0 ST Array 10818366 - -
          Affymetrix Rat Gene 1.1 ST Array 10818358 - -
          Rat Genome 230 2.0 Array 1393036_at - -
          Rat Genome 230 2.0 Array 1393036_PM_at - -
          Affymetrix HT Rat Focus Array 1393036_PM_at - -
          Affymetrix HT Rat Focus Array 1393036_at - -
          Affymetrix Rat Clariom S Assay PSR0200013745.rn.1 TC0200001990.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200013746.rn.1 TC0200001990.rn.1 RTA TC
          Rat Expression Set 230 1393036_PM_at - -
          Rat Expression Set 230 1393036_at - -
          Affymetrix Rat Gene 1.0 ST Array 10818358 - -
          Rat Genome U34 Set rc_AI044340_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21320237 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline