Search
Search

Mettl14| Gene Symbol: Mettl14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106470.1 | NP_001099940.1 | 5 - 6 | 518 | 97 | |
| XM_006233266.2 | XP_006233328.1 | 5 - 6 | 578 | 97 | ||
| GenBank mRNA | BC166789.1 | - | 5 - 6 | 424 | 7936402 | 97 |
Set Membership: |
|