Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01521982_m1
          See other MYBPHL GE Assays ›
          Gene Symbol:
          Mybphl
          Gene Name:
          myosin binding protein H-like
          Gene Aliases:
          RGD1310511
          Chromosome Location:
          Chr.2: 211158917 - 211173036 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01521982_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mybphl
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001014042.1 NP_001014064.1 8 - 9 1225 98
          GenBank mRNA BC087113.1 - 8 - 9 1225 7193587 98

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200001989.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mybphl

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P007163 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 489535 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 876887 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 162020 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 117193 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 487893 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 437419 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 899865 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 750161 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 529060 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 702589 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 662953 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 77500 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 159789 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P007163 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P007163 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P007163 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P778560 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 74992 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 6160 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10818354 - -
          Affymetrix Rat Gene 1.0 ST Array 10818355 - -
          Affymetrix Rat Gene 1.0 ST Array 10818351 - -
          Affymetrix Rat Gene 1.0 ST Array 10818349 - -
          Affymetrix Rat Gene 1.0 ST Array 10818357 - -
          Affymetrix Rat Gene 1.0 ST Array 10818352 - -
          Affymetrix Rat Gene 1.0 ST Array 10818356 - -
          Affymetrix Rat Gene 1.0 ST Array 10818350 - -
          Affymetrix Rat Gene 1.0 ST Array 10818353 - -
          Affymetrix Rat Gene 1.0 ST Array 10818348 - -
          Affymetrix Rat Gene 1.1 ST Array 10818347 - -
          Rat Genome 230 2.0 Array 1393036_at - -
          Rat Genome 230 2.0 Array 1378970_PM_at - -
          Rat Genome 230 2.0 Array 1393036_PM_at - -
          Rat Genome 230 2.0 Array 1389236_PM_at - -
          Rat Genome 230 2.0 Array 1378970_at - -
          Rat Genome 230 2.0 Array 1389236_at - -
          Affymetrix HT Rat Focus Array 1389236_PM_at - -
          Affymetrix HT Rat Focus Array 1378970_at - -
          Affymetrix HT Rat Focus Array 1378970_PM_at - -
          Affymetrix HT Rat Focus Array 1389236_at - -
          Affymetrix Rat Clariom S Assay PSR0200013725.rn.1 TC0200001989.rn.1 RTA TC
          Rat Expression Set 230 1389236_PM_at - -
          Rat Expression Set 230 1389236_at - -
          Rat Expression Set 230 1378970_at - -
          Rat Expression Set 230 1378970_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10818347 - -
          Rat Genome U34 Set rc_AI104915_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22184697 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1373006 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline