Search
Search
| Gene Symbol: Olfm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145777.1 | NP_665720.1 | 3 - 4 | 519 | 70 | |
| GenBank mRNA | AF442822.1 | - | 3 - 4 | 519 | 70 | |
| AF442823.1 | - | 410 | 70 | |||
Set Membership: |
|