Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01523036_m1
          See other ABCA4 GE Assays ›
          Gene Symbol:
          Abca4
          Gene Name:
          ATP-binding cassette, subfamily A (ABC1), member 4
          Gene Aliases:
          ABCR
          Chromosome Location:
          Chr.2: 225645455 - 225783288 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01523036_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Abca4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001107721.1 NP_001101191.1 49 - 50 6825 96
          XM_006233242.2 XP_006233304.1 49 - 50 7004 96

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200002112.rn.1

          Set Membership:

          Amplicon length between 86 and 100 Transporter 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          ATP-binding cassette (ABC) transporter primary active transporter transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          phospholipid transfer to membrane
          visual perception
          biological_process
          metabolic process
          phospholipid translocation
          photoreceptor cell maintenance
          transmembrane transport
          molecular_function
          phospholipid-translocating ATPase activity
          ATP binding
          phospholipid transporter activity
          ATPase activity, coupled to transmembrane movement of substances

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P116595 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 432420 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 818862 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 590152 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 747147 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 683727 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 895914 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 855729 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 955740 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 178243 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 137223 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 665430 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 184761 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 404615 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 746439 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 302771 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 971221 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 121383 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 199400 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 731540 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 191562 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 195499 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 598814 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 345510 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 910116 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 420153 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 91614 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 915391 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 110923 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 438496 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 894531 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 586816 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 463386 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 886491 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 588558 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 733735 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 556195 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 354957 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 338664 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 799887 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 54488 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 821806 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 688787 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 329227 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 24632 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 948593 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 229573 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 569302 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 681974 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 738483 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 568879 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 129985 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 497007 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 553496 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 683147 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 149103 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 465944 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 446825 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 359292 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 584422 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 657112 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 33485 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 342818 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 344314 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 580778 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 119505 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 538133 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 790829 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 189436 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 30585 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 802621 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 897487 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 541509 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 296396 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 171567 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 146626 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 186164 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 216954 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 646840 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 973402 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 221405 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 947554 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 207801 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 101430 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 893285 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 144673 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 920411 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 227939 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 698743 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 762321 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 506338 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 292871 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 643849 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 849439 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 969284 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 389082 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 763918 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 914749 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 187013 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 532785 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 330286 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 461665 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 740607 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 172972 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 49517 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 313341 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 424702 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 61098 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 120101 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 28261 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 115900 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 35761 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 162567 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 162201 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 138522 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 151007 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 130764 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 100043 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 104205 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 60773 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 19702 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 147020 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 81422 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P116596 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P116596 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P116596 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P22468 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P23003 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 94134 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 78150 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 100963 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10818757 - -
          Affymetrix Rat Gene 1.0 ST Array 10818764 - -
          Affymetrix Rat Gene 1.0 ST Array 10818768 - -
          Affymetrix Rat Gene 1.0 ST Array 10818743 - -
          Affymetrix Rat Gene 1.0 ST Array 10818750 - -
          Affymetrix Rat Gene 1.0 ST Array 10818783 - -
          Affymetrix Rat Gene 1.0 ST Array 10818745 - -
          Affymetrix Rat Gene 1.0 ST Array 10818754 - -
          Affymetrix Rat Gene 1.0 ST Array 10818761 - -
          Affymetrix Rat Gene 1.0 ST Array 10818767 - -
          Affymetrix Rat Gene 1.0 ST Array 10818786 - -
          Affymetrix Rat Gene 1.0 ST Array 10818772 - -
          Affymetrix Rat Gene 1.0 ST Array 10818785 - -
          Affymetrix Rat Gene 1.0 ST Array 10818744 - -
          Affymetrix Rat Gene 1.0 ST Array 10818791 - -
          Affymetrix Rat Gene 1.0 ST Array 10818773 - -
          Affymetrix Rat Gene 1.0 ST Array 10818747 - -
          Affymetrix Rat Gene 1.0 ST Array 10818775 - -
          Affymetrix Rat Gene 1.0 ST Array 10818762 - -
          Affymetrix Rat Gene 1.0 ST Array 10818756 - -
          Affymetrix Rat Gene 1.0 ST Array 10818787 - -
          Affymetrix Rat Gene 1.0 ST Array 10818782 - -
          Affymetrix Rat Gene 1.0 ST Array 10818779 - -
          Affymetrix Rat Gene 1.0 ST Array 10818759 - -
          Affymetrix Rat Gene 1.0 ST Array 10818758 - -
          Affymetrix Rat Gene 1.0 ST Array 10818788 - -
          Affymetrix Rat Gene 1.0 ST Array 10818771 - -
          Affymetrix Rat Gene 1.0 ST Array 10818790 - -
          Affymetrix Rat Gene 1.0 ST Array 10818765 - -
          Affymetrix Rat Gene 1.0 ST Array 10818749 - -
          Affymetrix Rat Gene 1.0 ST Array 10818755 - -
          Affymetrix Rat Gene 1.0 ST Array 10818748 - -
          Affymetrix Rat Gene 1.0 ST Array 10818774 - -
          Affymetrix Rat Gene 1.0 ST Array 10818746 - -
          Affymetrix Rat Gene 1.0 ST Array 10818781 - -
          Affymetrix Rat Gene 1.0 ST Array 10818753 - -
          Affymetrix Rat Gene 1.0 ST Array 10818769 - -
          Affymetrix Rat Gene 1.0 ST Array 10818752 - -
          Affymetrix Rat Gene 1.0 ST Array 10818777 - -
          Affymetrix Rat Gene 1.0 ST Array 10818792 - -
          Affymetrix Rat Gene 1.0 ST Array 10818780 - -
          Affymetrix Rat Gene 1.0 ST Array 10818766 - -
          Affymetrix Rat Gene 1.0 ST Array 10818789 - -
          Affymetrix Rat Gene 1.0 ST Array 10818784 - -
          Affymetrix Rat Gene 1.0 ST Array 10818770 - -
          Affymetrix Rat Gene 1.0 ST Array 10818778 - -
          Affymetrix Rat Gene 1.0 ST Array 10818776 - -
          Affymetrix Rat Gene 1.0 ST Array 10818763 - -
          Affymetrix Rat Gene 1.1 ST Array 10818742 - -
          Rat Genome 230 2.0 Array 1384603_PM_at - -
          Rat Genome 230 2.0 Array 1384603_at - -
          Affymetrix HT Rat Focus Array 1384603_PM_at - -
          Affymetrix HT Rat Focus Array 1384603_at - -
          Affymetrix Rat Clariom S Assay PSR0200014602.rn.1 TC0200002112.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200014601.rn.1 TC0200002112.rn.1 RTA TC
          Rat Expression Set 230 1384603_PM_at - -
          Rat Expression Set 230 1384603_at - -
          Affymetrix Rat Gene 1.0 ST Array 10818742 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20935489 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1356772 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline