Search
Search
| Gene Symbol: Lphn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF063102.1 | - | 7 - 8 | 1629 | 72 | |
| AF081148.1 | - | 8 - 9 | 2075 | 72 | ||
| AF081149.1 | - | 8 - 9 | 2075 | 72 | ||
| AF081150.1 | - | 8 - 9 | 2075 | 72 | ||
| AF081151.1 | - | 8 - 9 | 2075 | 72 | ||
| AF081152.1 | - | 8 - 9 | 2075 | 72 | ||
| AF081153.1 | - | 8 - 9 | 2075 | 72 | ||
Set Membership: |
|