Search
Search
| Gene Symbol: Glb1l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024358.1 | NP_001019529.2 | 12 - 13 | 1240 | 122 | |
| GenBank mRNA | BC083665.1 | - | 12 - 13 | 1240 | 7135489 | 122 |
Set Membership: |
|