Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01524960_m1
          See other HERPUD2 GE Assays ›
          Gene Symbol:
          Herpud2
          Gene Name:
          HERPUD family member 2
          Gene Aliases:
          RGD1307343
          Chromosome Location:
          Chr.8: 26266571 - 26311528 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01524960_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Herpud2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001024988.1 NP_001020159.1 6 - 7 1274 118
          XM_006242723.2 XP_006242785.1 6 - 7 1452 118
          XM_006242724.2 XP_006242786.1 6 - 7 1514 118
          GenBank mRNA BC081861.1 - 6 - 7 1274 7134757 118

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800002117.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Herpud2

          Gene Ontology Categories:

          Function(s)

          response to unfolded protein
          spermatogenesis

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P593381 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 356683 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 428455 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 40561 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 97877 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 568426 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 705456 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 56224 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 945519 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 314576 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 79533 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 390230 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 922582 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 704313 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 227615 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 311951 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 812297 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 316502 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 749437 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 282693 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 240074 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 800870 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 187116 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 23280 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 285035 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 368402 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 330580 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 967333 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 182255 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 654470 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 219910 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 835415 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 648825 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 618354 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 921196 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 762105 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 177762 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 146588 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 157427 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 62824 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 22140 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 56697 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P593381 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P593381 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P593381 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P593381 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 16906 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 2090 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 48081 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 30054 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10915833 - -
          Affymetrix Rat Gene 1.0 ST Array 10915829 - -
          Affymetrix Rat Gene 1.0 ST Array 10915834 - -
          Affymetrix Rat Gene 1.0 ST Array 10915835 - -
          Affymetrix Rat Gene 1.0 ST Array 10915836 - -
          Affymetrix Rat Gene 1.0 ST Array 10915828 - -
          Affymetrix Rat Gene 1.0 ST Array 10915830 - -
          Affymetrix Rat Gene 1.0 ST Array 10915827 - -
          Affymetrix Rat Gene 1.0 ST Array 10915832 - -
          Affymetrix Rat Gene 1.1 ST Array 10915826 - -
          Rat Genome 230 2.0 Array 1377613_PM_at - -
          Rat Genome 230 2.0 Array 1377613_at - -
          Affymetrix HT Rat Focus Array 1377613_at - -
          Affymetrix HT Rat Focus Array 1377613_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0800017294.rn.1 TC0800002117.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800017299.rn.1 TC0800002117.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800017301.rn.1 TC0800002117.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800017295.rn.1 TC0800002117.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800017297.rn.1 TC0800002117.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800017296.rn.1 TC0800002117.rn.1 RTA TC
          Rat Expression Set 230 1377613_at - -
          Rat Expression Set 230 1377613_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10915826 - -
          Rat Genome U34 Set rc_AI235382_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20998073 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1351451 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1650292 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline