Search
Search

Prickle3| Gene Symbol: Prickle3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014110.1 | NP_001014132.1 | 2 - 3 | 195 | 116 | |
| GenBank mRNA | BC085918.1 | - | 2 - 3 | 195 | 7108577 | 116 |
Set Membership: |
|