Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01525445_m1
          See other RAI14 GE Assays ›
          Gene Symbol:
          Rai14
          Gene Name:
          retinoic acid induced 14
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 60546083 - 60684376 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01525445_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rai14
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001011947.1 NP_001011947.1 16 - 17 2873 83
          XM_006232060.2 XP_006232122.1 17 - 18 3447 83
          XM_006232061.2 XP_006232123.1 17 - 18 3020 83
          XM_006232062.2 XP_006232124.1 17 - 18 3885 83
          XM_006232063.2 XP_006232125.1 16 - 17 3360 83
          XM_006232064.2 XP_006232126.1 15 - 16 3273 83
          XM_008760765.1 XP_008758987.1 15 - 16 3816 83
          GenBank mRNA BC085775.1 - 16 - 17 2873 7096978 83

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200003073.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rai14

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P091388 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 779928 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 766176 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 799195 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 793757 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 384345 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 722085 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 651446 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 257856 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 757967 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 461794 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 47001 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 14939 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 427893 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 498219 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 289406 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 633408 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 544615 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 378632 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 507719 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 370876 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 544656 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 4527 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 582857 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 660630 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 368230 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 460255 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 143072 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 230096 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 699384 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 627257 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 588508 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 446254 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 18932 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 566160 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 304771 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 102121 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 725637 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 196813 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 230799 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 655174 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 782261 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 522826 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 88283 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 198546 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 285367 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 634875 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 401326 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 951591 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 614112 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 768876 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 593027 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 730214 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 422630 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 275770 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 444061 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 887258 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 528087 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 730975 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 831227 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 237280 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 155768 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 920313 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 671645 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 876744 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 569185 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 458966 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 277 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 867757 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 273364 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 259988 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 47278 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 386966 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 730176 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 631481 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 852420 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 94373 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 439502 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 105860 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 523889 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 873805 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 338609 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 159347 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 597136 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 895629 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 289824 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 348857 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 479893 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 549300 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 341671 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 112572 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 789496 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 274653 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 648963 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 849062 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 640260 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 10293 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 290319 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 363941 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 593982 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 381784 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 739902 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 419664 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 742904 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 861783 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 112719 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 266972 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 70455 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 240619 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 932026 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 354172 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 143554 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 117324 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 93669 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 29719 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 106713 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 19704 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 22256 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 86764 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 64007 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 55509 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 113413 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 58066 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 142301 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 147360 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 61795 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 87810 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 108166 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P091388 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P091388 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P091388 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P20995 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 56780 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 45658 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 89340 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 100943 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 92255 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 1544 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 71934 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 86377 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10821929 - -
          Affymetrix Rat Gene 1.0 ST Array 10821939 - -
          Affymetrix Rat Gene 1.0 ST Array 10821935 - -
          Affymetrix Rat Gene 1.0 ST Array 10821930 - -
          Affymetrix Rat Gene 1.0 ST Array 10821931 - -
          Affymetrix Rat Gene 1.0 ST Array 10821938 - -
          Affymetrix Rat Gene 1.0 ST Array 10821927 - -
          Affymetrix Rat Gene 1.0 ST Array 10821933 - -
          Affymetrix Rat Gene 1.0 ST Array 10821928 - -
          Affymetrix Rat Gene 1.0 ST Array 10821934 - -
          Affymetrix Rat Gene 1.0 ST Array 10821942 - -
          Affymetrix Rat Gene 1.0 ST Array 10821937 - -
          Affymetrix Rat Gene 1.0 ST Array 10821941 - -
          Affymetrix Rat Gene 1.0 ST Array 10821936 - -
          Affymetrix Rat Gene 1.0 ST Array 10821925 - -
          Affymetrix Rat Gene 1.0 ST Array 10821926 - -
          Affymetrix Rat Gene 1.1 ST Array 10821924 - -
          Rat Genome 230 2.0 Array 1384528_at - -
          Rat Genome 230 2.0 Array 1389263_at - -
          Rat Genome 230 2.0 Array 1384528_PM_at - -
          Rat Genome 230 2.0 Array 1389263_PM_at - -
          Affymetrix HT Rat Focus Array 1384528_at - -
          Affymetrix HT Rat Focus Array 1384528_PM_at - -
          Affymetrix HT Rat Focus Array 1389263_PM_at - -
          Affymetrix HT Rat Focus Array 1389263_at - -
          Affymetrix Rat Clariom S Assay PSR0200021523.rn.1 TC0200003073.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200021522.rn.1 TC0200003073.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200021521.rn.1 TC0200003073.rn.1 RTA TC
          Rat Expression Set 230 1389263_at - -
          Rat Expression Set 230 1389263_PM_at - -
          Rat Expression Set 230 1384528_at - -
          Rat Expression Set 230 1384528_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10821924 - -
          Rat Genome U34 Set rc_AI234852_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21316229 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1375727 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376614 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline