Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ddah2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001165936.1 | NP_001159408.1 | 5 - 6 | 787 | 105 | |
NM_212532.2 | NP_997697.1 | 5 - 6 | 834 | 105 | ||
XM_008772714.1 | XP_008770936.1 | 6 - 7 | 933 | 105 | ||
GenBank mRNA | BC086443.1 | - | 5 - 6 | 735 | 7303579 | 105 |
BC097930.1 | - | 5 - 6 | 824 | 7388716 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons |