Search
Search

Zdhhc14| Gene Symbol: Zdhhc14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039343.1 | NP_001034432.1 | 4 - 5 | 1585 | 93 | |
| XM_006227884.2 | XP_006227946.1 | 4 - 5 | 1548 | 93 | ||
| XM_006227885.2 | XP_006227947.1 | 4 - 5 | 1553 | 93 | ||
| XM_006227886.2 | XP_006227948.1 | 4 - 5 | 1552 | 93 | ||
| GenBank mRNA | AY886530.1 | - | 4 - 5 | 1585 | 93 | |
Set Membership: |
|