Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01526929_g1
          See other ANKIB1 GE Assays ›
          Gene Symbol:
          Ankib1
          Gene Name:
          ankyrin repeat and IBR domain containing 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 27473225 - 27597206 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01526929_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ankib1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001134781.1 NP_001128253.1 19 - 20 2833 70
          XM_006236049.2 XP_006236111.1 19 - 20 3103 70
          XM_006236050.2 XP_006236112.1 20 - 21 3957 70
          XM_006236051.2 XP_006236113.1 19 - 20 2553 70
          XM_008762709.1 XP_008760931.1 19 - 20 4075 70
          XM_008762710.1 XP_008760932.1 20 - 21 3469 70
          XM_008762711.1 XP_008760933.1 19 - 20 4057 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400000290.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase

          Gene Ontology Categories:

          Process(es)

          zinc ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P043579 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P043574 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 386638 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 934492 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 417600 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 457812 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 665420 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 273589 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 793731 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 520577 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 933867 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 284169 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 119968 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 219001 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 400622 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 196024 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 130772 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 510598 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 42695 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 141933 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 119013 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 776109 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 159625 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 728425 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 194293 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 886363 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 356212 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 688163 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 327742 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 785715 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 241958 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 965561 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 619341 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 571295 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 509359 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 756176 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 658185 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 252903 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 262729 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 825538 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 59212 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 494491 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 938974 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 919471 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 220653 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 173420 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 610516 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 513065 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 949221 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 761688 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 189458 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 506927 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 482344 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 464330 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 90781 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 567629 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 471657 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 61570 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 383500 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 742182 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 329021 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 523456 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 133358 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 182908 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 716363 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 122953 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 373174 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 316932 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 348780 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 460538 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 269593 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 774440 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 156381 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 456468 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 664911 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 954097 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 22589 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 831649 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843751 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 804790 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 824965 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 128021 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 948661 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 108326 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 74931 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 163411 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 156323 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 172665 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 124185 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 81559 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 15988 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 158863 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 94592 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 108687 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 38823 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 46984 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P043574 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P043579 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P043579 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P043574 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P043579 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P043574 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P462543 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 73637 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 93518 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10853498 - -
          Affymetrix Rat Gene 1.0 ST Array 10853496 - -
          Affymetrix Rat Gene 1.0 ST Array 10853497 - -
          Affymetrix Rat Gene 1.0 ST Array 10853486 - -
          Affymetrix Rat Gene 1.0 ST Array 10853493 - -
          Affymetrix Rat Gene 1.0 ST Array 10853489 - -
          Affymetrix Rat Gene 1.0 ST Array 10853484 - -
          Affymetrix Rat Gene 1.0 ST Array 10853485 - -
          Affymetrix Rat Gene 1.0 ST Array 10853490 - -
          Affymetrix Rat Gene 1.0 ST Array 10853491 - -
          Affymetrix Rat Gene 1.0 ST Array 10853487 - -
          Affymetrix Rat Gene 1.0 ST Array 10853499 - -
          Affymetrix Rat Gene 1.0 ST Array 10853500 - -
          Affymetrix Rat Gene 1.0 ST Array 10853482 - -
          Affymetrix Rat Gene 1.0 ST Array 10853488 - -
          Affymetrix Rat Gene 1.0 ST Array 10853492 - -
          Affymetrix Rat Gene 1.0 ST Array 10853494 - -
          Affymetrix Rat Gene 1.0 ST Array 10853483 - -
          Affymetrix Rat Gene 1.0 ST Array 10853495 - -
          Affymetrix Rat Gene 1.1 ST Array 10853481 - -
          Rat Genome 230 2.0 Array 1397825_at - -
          Rat Genome 230 2.0 Array 1397825_PM_at - -
          Rat Genome 230 2.0 Array 1376357_PM_at - -
          Rat Genome 230 2.0 Array 1381586_PM_at - -
          Rat Genome 230 2.0 Array 1376357_at - -
          Rat Genome 230 2.0 Array 1381586_at - -
          Affymetrix HT Rat Focus Array 1397825_PM_at - -
          Affymetrix HT Rat Focus Array 1376357_at - -
          Affymetrix HT Rat Focus Array 1376357_PM_at - -
          Affymetrix HT Rat Focus Array 1381586_PM_at - -
          Affymetrix HT Rat Focus Array 1397825_at - -
          Affymetrix HT Rat Focus Array 1381586_at - -
          Affymetrix Rat Clariom S Assay PSR0400002084.rn.1 TC0400000290.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400002086.rn.1 TC0400000290.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400002085.rn.1 TC0400000290.rn.1 RTA TC
          Rat Expression Set 230 1376357_PM_at - -
          Rat Expression Set 230 1376357_at - -
          Rat Expression Set 230 1397825_at - -
          Rat Expression Set 230 1397825_PM_at - -
          Rat Expression Set 230 1381586_at - -
          Rat Expression Set 230 1381586_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10853481 - -
          Rat Genome U34 Set rc_AA875143_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20711332 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1374395 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline