Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01527590_m1
          See other CRB3 GE Assays ›
          Gene Symbol:
          Crb3
          Gene Name:
          crumbs family member 3
          Gene Aliases:
          -
          Chromosome Location:
          Chr.9: 9982529 - 9986892 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01527590_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Crb3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001025661.1 NP_001020832.1 3 - 4 292 107
          XM_006244303.2 XP_006244365.1 3 - 4 1409 107
          GenBank mRNA BC097382.1 - 3 - 4 292 7446098 107
          FQ223867.1 - 525 107

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0900001452.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Crb3

          Panther Classification:

          Molecular Function -

          cell adhesion molecule defense/immunity protein immunoglobulin receptor superfamily

          Gene Ontology Categories:

          Function(s) Process(es)

          protein localization to plasma membrane
          SH3 domain binding
          protein domain specific binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1043302 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1043302 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1043302 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1043302 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1043302 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 81502 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10931340 - -
          Affymetrix Rat Gene 1.0 ST Array 10931339 - -
          Affymetrix Rat Gene 1.0 ST Array 10931338 - -
          Affymetrix Rat Gene 1.0 ST Array 10931337 - -
          Affymetrix Rat Gene 1.1 ST Array 10931336 - -
          Rat Genome 230 2.0 Array 1383648_PM_x_at - -
          Rat Genome 230 2.0 Array 1383647_a_at - -
          Rat Genome 230 2.0 Array 1383647_PM_a_at - -
          Rat Genome 230 2.0 Array 1383648_x_at - -
          Rat Genome 230 2.0 Array 1384397_PM_x_at - -
          Rat Genome 230 2.0 Array 1384531_at - -
          Rat Genome 230 2.0 Array 1384531_PM_at - -
          Rat Genome 230 2.0 Array 1384397_x_at - -
          Affymetrix HT Rat Focus Array 1383647_a_at - -
          Affymetrix HT Rat Focus Array 1384397_PM_x_at - -
          Affymetrix HT Rat Focus Array 1384397_x_at - -
          Affymetrix HT Rat Focus Array 1384531_PM_at - -
          Affymetrix HT Rat Focus Array 1383648_PM_x_at - -
          Affymetrix HT Rat Focus Array 1383647_PM_a_at - -
          Affymetrix HT Rat Focus Array 1383648_x_at - -
          Affymetrix HT Rat Focus Array 1384531_at - -
          Affymetrix Rat Clariom S Assay PSR0900011976.rn.1 TC0900001452.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900011977.rn.1 TC0900001452.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900011978.rn.1 TC0900001452.rn.1 RTA TC
          Rat Expression Set 230 1384397_PM_x_at - -
          Rat Expression Set 230 1383647_PM_a_at - -
          Rat Expression Set 230 1384531_PM_at - -
          Rat Expression Set 230 1384397_x_at - -
          Rat Expression Set 230 1383648_PM_x_at - -
          Rat Expression Set 230 1383648_x_at - -
          Rat Expression Set 230 1383647_a_at - -
          Rat Expression Set 230 1384531_at - -
          Affymetrix Rat Gene 1.0 ST Array 10931336 - -
          Rat Genome U34 Set rc_AI104432_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21539122 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1374618 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-d4j86:80/100.66.77.21:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline