Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01528076_m1
          See other GPC2 GE Assays ›
          Gene Symbol:
          Gpc2
          Gene Name:
          glypican 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.12: 19593041 - 19599374 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01528076_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gpc2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_138511.1 NP_612520.1 9 - 10 1713 86
          GenBank mRNA L20468.1 - 9 - 10 1713 86

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1200001258.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gpc2

          Gene Ontology Categories:

          Function(s) Process(es)

          smoothened signaling pathway
          neuron differentiation
          molecular_function
          heparan sulfate proteoglycan binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P685404 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 72159 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 380168 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 929127 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 657837 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 18889 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 34551 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P059257 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P059257 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P059257 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P685404 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 35046 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10760900 - -
          Affymetrix Rat Gene 1.0 ST Array 10760898 - -
          Affymetrix Rat Gene 1.0 ST Array 10760897 - -
          Affymetrix Rat Gene 1.0 ST Array 10760905 - -
          Affymetrix Rat Gene 1.0 ST Array 10760896 - -
          Affymetrix Rat Gene 1.0 ST Array 10760903 - -
          Affymetrix Rat Gene 1.0 ST Array 10760899 - -
          Affymetrix Rat Gene 1.0 ST Array 10760904 - -
          Affymetrix Rat Gene 1.0 ST Array 10760902 - -
          Affymetrix Rat Gene 1.0 ST Array 10760901 - -
          Affymetrix Rat Gene 1.1 ST Array 10760895 - -
          Rat Genome 230 2.0 Array 1370448_at - -
          Rat Genome 230 2.0 Array 1370448_PM_at - -
          Affymetrix HT Rat Focus Array 1370448_PM_at - -
          Affymetrix HT Rat Focus Array 1370448_at - -
          Affymetrix Rat Clariom S Assay PSR1200010635.rn.1 TC1200001258.rn.1 RTA TC
          Rat Expression Set 230 1370448_PM_at - -
          Rat Expression Set 230 1370448_at - -
          Affymetrix Rat Gene 1.0 ST Array 10760895 - -
          Rat Genome U34 Set L20468_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21875887 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline