Search
Search

Suv39h1| Gene Symbol: Suv39h1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106956.1 | NP_001100426.1 | 2 - 3 | 617 | 142 | |
| XM_006256703.2 | XP_006256765.1 | 2 - 3 | 833 | 142 | ||
| XM_006256704.2 | XP_006256766.1 | 2 - 3 | 409 | 142 | ||
| XM_006256705.2 | XP_006256767.1 | 2 - 3 | 410 | 142 | ||
| GenBank mRNA | BC160871.1 | - | 2 - 3 | 184 | 7364965 | 142 |
Set Membership: |
|