Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01528326_m1
          See other OTUD5 GE Assays ›
          Gene Symbol:
          Otud5
          Gene Name:
          OTU deubiquitinase 5
          Gene Aliases:
          -
          Chromosome Location:
          Chr.X: 15470970 - 15504374 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01528326_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Otud5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001037496.1 NP_001032585.1 7 - 8 1240 95
          XM_006256737.2 XP_006256799.1 7 - 8 1890 95
          XM_006256738.2 XP_006256800.1 7 - 8 1889 95
          XM_006256739.2 XP_006256801.1 7 - 8 1888 95
          XM_006256740.2 XP_006256802.1 8 - 9 1760 95
          XM_006256741.2 XP_006256803.1 8 - 9 1759 95
          GenBank mRNA BC110054.1 - 7 - 8 1240 7446041 95

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0X00001496.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Otud5

          Panther Classification:

          Molecular Function -

          cysteine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          response to lipopolysaccharide
          protein K63-linked deubiquitination
          protein K48-linked deubiquitination
          thiol-dependent ubiquitin-specific protease activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P475932 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 912549 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 567369 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 937212 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 18055 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 486966 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 653541 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 906041 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 631623 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 861084 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 237098 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 169931 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 851450 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 310329 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 796307 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 541833 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 820897 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 159901 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 275372 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 220018 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 681404 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 792279 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 428053 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 409503 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 294754 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 82721 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 126643 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 6356 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 161137 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 53976 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P475932 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P475932 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P475932 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P475932 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 88632 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 104640 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 58352 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 104581 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10937001 - -
          Affymetrix Rat Gene 1.0 ST Array 10937008 - -
          Affymetrix Rat Gene 1.0 ST Array 10937012 - -
          Affymetrix Rat Gene 1.0 ST Array 10937002 - -
          Affymetrix Rat Gene 1.0 ST Array 10937003 - -
          Affymetrix Rat Gene 1.0 ST Array 10937004 - -
          Affymetrix Rat Gene 1.0 ST Array 10937011 - -
          Affymetrix Rat Gene 1.0 ST Array 10937010 - -
          Affymetrix Rat Gene 1.0 ST Array 10937006 - -
          Affymetrix Rat Gene 1.0 ST Array 10937005 - -
          Affymetrix Rat Gene 1.0 ST Array 10937009 - -
          Affymetrix Rat Gene 1.0 ST Array 10937007 - -
          Affymetrix Rat Gene 1.1 ST Array 10937000 - -
          Rat Genome 230 2.0 Array 1373231_PM_at - -
          Rat Genome 230 2.0 Array 1373231_at - -
          Affymetrix HT Rat Focus Array 1373231_at - -
          Affymetrix HT Rat Focus Array 1373231_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0X00009499.rn.1 TC0X00001496.rn.1 RTA TC
          Rat Expression Set 230 1373231_PM_at - -
          Rat Expression Set 230 1373231_at - -
          Affymetrix Rat Gene 1.0 ST Array 10937000 - -
          Rat Genome U34 Set rc_AA998154_at - -
          Rat Genome U34 Set rc_AI145368_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1362346 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline