Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01528818_g1
          See other ANGPTL4 GE Assays ›
          Gene Symbol:
          Angptl4
          Gene Name:
          angiopoietin-like 4
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 18627814 - 18634043 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01528818_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Angptl4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_199115.2 NP_954546.1 5 - 6 898 113
          GenBank mRNA AY393999.1 - 5 - 6 758 113
          BC078944.1 - 5 - 6 898 7109431 113
          FQ218836.1 - 2 - 3 168 113

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700002295.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Rat Hypoxia Panel 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Angptl4

          Panther Classification:

          Molecular Function -

          intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          response to hypoxia
          regulation of lipid metabolic process
          cell differentiation
          negative regulation of apoptotic process
          negative regulation of catalytic activity
          negative regulation of lipoprotein lipase activity
          protein oligomerization
          protein homooligomerization
          triglyceride homeostasis
          negative regulation of endothelial cell apoptotic process
          enzyme inhibitor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P088382 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 800570 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 282258 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 700248 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 911621 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P088382 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P088382 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P088382 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P391057 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 52908 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10901172 - -
          Affymetrix Rat Gene 1.0 ST Array 10901170 - -
          Affymetrix Rat Gene 1.0 ST Array 10901175 - -
          Affymetrix Rat Gene 1.0 ST Array 10901174 - -
          Affymetrix Rat Gene 1.0 ST Array 10901171 - -
          Affymetrix Rat Gene 1.0 ST Array 10901167 - -
          Affymetrix Rat Gene 1.0 ST Array 10901173 - -
          Affymetrix Rat Gene 1.0 ST Array 10901168 - -
          Affymetrix Rat Gene 1.0 ST Array 10901169 - -
          Affymetrix Rat Gene 1.1 ST Array 10901166 - -
          Rat Genome 230 2.0 Array 1388924_at - -
          Rat Genome 230 2.0 Array 1388924_PM_at - -
          Affymetrix HT Rat Focus Array 1388924_PM_at - -
          Affymetrix HT Rat Focus Array 1388924_at - -
          Affymetrix Rat Clariom S Assay PSR0700018406.rn.1 TC0700002295.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700018408.rn.1 TC0700002295.rn.1 RTA TC
          Rat Expression Set 230 1388924_at - -
          Rat Expression Set 230 1388924_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10901166 - -
          Rat Genome U34 Set rc_AA944481_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21515889 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1530352 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1360466 - -
          RatRef-12 Expression BeadChip ILMN_161135 - -
          RatRef-12 Expression BeadChip ILMN_68167 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline