Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01529747_g1
          See other UBL4A GE Assays ›
          Gene Symbol:
          Ubl4a
          Gene Name:
          ubiquitin-like 4A
          Gene Aliases:
          RGD1563983, Ubl4
          Chromosome Location:
          Chr.X: 156340919 - 156343771 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01529747_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ubl4a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106346.2 NP_001099816.1 1 - 2 122 86
          GenBank mRNA BC166513.1 - 1 - 2 122 7129152 86

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0X00001293.rn.1

          Set Membership:

          Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s)

          transport
          tail-anchored membrane protein insertion into ER membrane

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1040230 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 132685 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 807123 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 27639 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1040230 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1040230 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1040230 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1040230 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10936069 - -
          Affymetrix Rat Gene 1.0 ST Array 10936070 - -
          Affymetrix Rat Gene 1.0 ST Array 10936071 - -
          Affymetrix Rat Gene 1.0 ST Array 10933603 - -
          Affymetrix Rat Gene 1.1 ST Array 10936067 - -
          Affymetrix Rat Gene 1.1 ST Array 10933601 - -
          Rat Genome 230 2.0 Array 1392906_PM_at - -
          Rat Genome 230 2.0 Array 1392906_at - -
          Rat Genome 230 2.0 Array 1373249_PM_at - -
          Rat Genome 230 2.0 Array 1373249_at - -
          Affymetrix HT Rat Focus Array 1392906_PM_at - -
          Affymetrix HT Rat Focus Array 1373249_at - -
          Affymetrix HT Rat Focus Array 1392906_at - -
          Affymetrix HT Rat Focus Array 1373249_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0X00007677.rn.1 TC0X00001293.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00007679.rn.1 TC0X00001293.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00007675.rn.1 TC0X00001293.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00007676.rn.1 TC0X00001293.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00007678.rn.1 TC0X00001293.rn.1 RTA TC
          Rat Expression Set 230 1373249_at - -
          Rat Expression Set 230 1373249_PM_at - -
          Rat Expression Set 230 1392906_PM_at - -
          Rat Expression Set 230 1392906_at - -
          Affymetrix Rat Gene 1.0 ST Array 10936067 - -
          Rat Genome U34 Set rc_AA817861_at - -
          Rat Genome U34 Set rc_AI232649_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21532376 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1372555 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline