Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01530256_g1
          See other LOC498750 GE Assays ›
          Gene Symbol:
          LOC498750 LOC100911664
          Gene Name:
          similar to cDNA sequence BC005537
          uncharacterized LOC100911664
          Gene Aliases:
          -
          -
          Chromosome Location:
          Chr.3: 17979714 - 17980319 on Build Rnor_6.0
          Chromosome Location:
          Chr.17: 42256971 - 42268996 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn01530256_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC498750
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001017510.3 NP_001017510.1 1 601 100
          GenBank mRNA AY325226.1 - 1 601 100
          BC085940.1 - 4 - 5 1062 7123624 100
          BC089212.1 - 4 - 5 860 7302835 100
          Gene Symbol: LOC100911664
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_003751691.3 XP_003751739.1 4 - 5 2777 100

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene LOC498750
          Click here to find siRNAs for Gene LOC100911664

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P337253 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 780175 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 372590 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 678968 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 368305 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 530015 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 644630 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 536303 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 682320 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 416403 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 131697 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 667223 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 511650 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 244852 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 513494 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 375341 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 471424 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 83472 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 31311 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P337253 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P337253 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P337253 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P337253 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 71100 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 70103 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 22886 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10798389 - -
          Affymetrix Rat Gene 1.0 ST Array 10856188 - -
          Affymetrix Rat Gene 1.0 ST Array 10798388 - -
          Affymetrix Rat Gene 1.0 ST Array 10798387 - -
          Affymetrix Rat Gene 1.1 ST Array 10798386 - -
          Affymetrix Rat Gene 1.1 ST Array 10856187 - -
          Rat Genome 230 2.0 Array 1372711_PM_at - -
          Rat Genome 230 2.0 Array 1372711_at - -
          Affymetrix HT Rat Focus Array 1372711_at - -
          Affymetrix HT Rat Focus Array 1372711_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1700012099.rn.1 TC1700001770.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700012096.rn.1 TC1700001770.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700012098.rn.1 TC1700001770.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700012097.rn.1 TC1700001770.rn.1 RTA TC
          Rat Expression Set 230 1372711_at - -
          Rat Expression Set 230 1372711_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10856187 - -
          Affymetrix Rat Gene 1.0 ST Array 10798386 - -
          Rat Genome U34 Set rc_AA851214_at - -
          Rat Genome U34 Set rc_AA819362_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20723479 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1355534 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline