Search
Search
| Gene Symbol: Gata4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144730.1 | NP_653331.1 | 3 - 4 | 1343 | 85 | |
| XM_006252189.2 | XP_006252251.1 | 2 - 3 | 923 | 85 | ||
| GenBank mRNA | L22761.1 | - | 3 - 4 | 1343 | 85 | |
Set Membership: |
|