Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01530621_m1
          See other CCPG1 GE Assays ›
          Gene Symbol:
          Ccpg1
          Gene Name:
          cell cycle progression 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.8: 79660634 - 79692044 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01530621_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ccpg1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001108770.1 NP_001102240.1 8 - 9 1981 120
          XM_006243341.2 XP_006243403.1 8 - 9 2010 120
          XM_008766320.1 XP_008764542.1 10 - 11 2094 120

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800001081.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Rattus norvegicus Probe spans exons

          Gene Ontology Categories:

          Function(s)

          positive regulation of cell proliferation
          positive regulation of cell cycle
          positive regulation of transcription from RNA polymerase II promoter
          regulation of Rho guanyl-nucleotide exchange factor activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P019135 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P019130 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 727188 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 791597 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 82227 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 643643 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 539503 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 757208 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 591604 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 352513 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 764353 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 491139 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 136171 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 127686 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 502733 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 21281 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 709323 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 391837 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 869418 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 420750 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 643449 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 960702 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 584174 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 296666 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 834978 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 307384 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 307308 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 107007 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 140208 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 4337 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 63418 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P019135 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P019130 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P019135 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P019130 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P019135 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P019130 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P309629 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P714633 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 94667 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 83421 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10911519 - -
          Affymetrix Rat Gene 1.0 ST Array 10911515 - -
          Affymetrix Rat Gene 1.0 ST Array 10911521 - -
          Affymetrix Rat Gene 1.0 ST Array 10918658 - -
          Affymetrix Rat Gene 1.0 ST Array 10911513 - -
          Affymetrix Rat Gene 1.0 ST Array 10911511 - -
          Affymetrix Rat Gene 1.0 ST Array 10911514 - -
          Affymetrix Rat Gene 1.0 ST Array 10911520 - -
          Affymetrix Rat Gene 1.0 ST Array 10911517 - -
          Affymetrix Rat Gene 1.0 ST Array 10911512 - -
          Affymetrix Rat Gene 1.0 ST Array 10911516 - -
          Affymetrix Rat Gene 1.1 ST Array 10918657 - -
          Affymetrix Rat Gene 1.1 ST Array 10911510 - -
          Rat Genome 230 2.0 Array 1375027_PM_at - -
          Rat Genome 230 2.0 Array 1396317_at - -
          Rat Genome 230 2.0 Array 1375027_at - -
          Rat Genome 230 2.0 Array 1396317_PM_at - -
          Affymetrix HT Rat Focus Array 1396317_PM_at - -
          Affymetrix HT Rat Focus Array 1396317_at - -
          Affymetrix Rat Clariom S Assay PSR0800007924.rn.1 TC0800001081.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800007921.rn.1 TC0800001081.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800007922.rn.1 TC0800001081.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800007920.rn.1 TC0800001081.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800007919.rn.1 TC0800001081.rn.1 RTA TC
          Rat Expression Set 230 1375027_at - -
          Rat Expression Set 230 1375027_PM_at - -
          Rat Expression Set 230 1396317_at - -
          Rat Expression Set 230 1396317_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10911510 - -
          Rat Genome U34 Set rc_AI029180_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1352750 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1371005 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline