Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01531989_g1
          See other IER3 GE Assays ›
          Gene Symbol:
          Ier3
          Gene Name:
          immediate early response 3
          Gene Aliases:
          Dif2, Iex1, Ler3, Prg1
          Chromosome Location:
          Chr.20: 3438798 - 3440002 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01531989_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ier3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_212505.2 NP_997670.1 1 - 2 246 55
          GenBank mRNA AW916683.1 - 1 - 2 247 55
          BC099831.1 - 1 - 2 222 7933804 55
          FM066628.1 - 1 - 2 231 55

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC2000001040.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ier3

          Gene Ontology Categories:

          Function(s)

          response to protozoan
          negative regulation of systemic arterial blood pressure
          regulation of DNA repair
          mitotic G2 DNA damage checkpoint
          intrinsic apoptotic signaling pathway in response to DNA damage
          positive regulation of protein catabolic process
          negative regulation of glycolytic process
          regulation of nucleocytoplasmic transport
          negative regulation of inflammatory response
          negative regulation of mitochondrial outer membrane permeabilization involved in apoptotic signaling pathway
          regulation of reactive oxygen species metabolic process
          regulation of response to DNA damage stimulus

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P515405 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 396993 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 23525 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P515405 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P515405 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P515405 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P515405 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 84278 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10831080 - -
          Affymetrix Rat Gene 1.0 ST Array 10831079 - -
          Affymetrix Rat Gene 1.0 ST Array 10831078 - -
          Affymetrix Rat Gene 1.1 ST Array 10831077 - -
          Rat Genome 230 2.0 Array 1388587_PM_at - -
          Rat Genome 230 2.0 Array 1388587_at - -
          Affymetrix HT Rat Focus Array 1388587_PM_at - -
          Affymetrix HT Rat Focus Array 1388587_at - -
          Affymetrix Rat Clariom S Assay PSR2000008438.rn.1 TC2000001040.rn.1 RTA TC
          Rat Expression Set 230 1388587_at - -
          Rat Expression Set 230 1388587_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10831077 - -
          Rat Genome U34 Set X96437mRNA_at - -
          Rat Genome U34 Set X96437mRNA_g_at - -
          Rat Genome U34 Set rc_AI169177_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21590666 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1371962 - -
          RatRef-12 Expression BeadChip ILMN_56245 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline