Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01532757_g1
          See other KRT8 GE Assays ›
          Gene Symbol:
          Krt8
          Gene Name:
          keratin 8, type II
          Gene Aliases:
          CYKER, Krt2-8
          Chromosome Location:
          Chr.7: 143596511 - 143603745 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01532757_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Krt8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_199370.1 NP_955402.1 8 - 9 1259 114
          GenBank mRNA AY464139.1 - 8 - 9 1259 114
          BC091106.1 - 8 - 9 1317 7381703 114
          BC097497.1 - 8 - 9 1282 7462062 114
          M63482.1 - 8 - 9 1323 114
          S76054.1 - 8 - 9 1323 114

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Krt8

          Gene Ontology Categories:

          Function(s) Process(es)

          tumor necrosis factor-mediated signaling pathway
          sarcomere organization
          response to hydrostatic pressure
          response to other organism
          cell differentiation involved in embryonic placenta development
          extrinsic apoptotic signaling pathway
          hepatocyte apoptotic process
          structural molecule activity
          protein binding
          protein complex binding
          scaffold protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P784063 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 630634 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 728671 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 252701 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 357796 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 77090 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P784063 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P138355 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P138355 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P784063 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P784063 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P138355 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P784063 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 702134683 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700537172 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 31524 - -
          Agilent Rat cDNA Microarray (G4105A) 700537172 - -
          Agilent Rat cDNA Microarray (G4105A) 702134683 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10907525 - -
          Affymetrix Rat Gene 1.0 ST Array 10907527 - -
          Affymetrix Rat Gene 1.0 ST Array 10907528 - -
          Affymetrix Rat Gene 1.0 ST Array 10907526 - -
          Affymetrix Rat Gene 1.0 ST Array 10751353 - -
          Affymetrix Rat Gene 1.1 ST Array 10907524 - -
          Affymetrix Rat Gene 1.1 ST Array 10751352 - -
          Rat Genome 230 2.0 Array 1371530_at - -
          Rat Genome 230 2.0 Array 1371530_PM_at - -
          Affymetrix HT Rat Focus Array 1371530_at - -
          Affymetrix HT Rat Focus Array 1371530_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0700031973.rn.1 TC0700003958.rn.1 RTA TC
          Rat Expression Set 230 1371530_PM_at - -
          Rat Expression Set 230 1371530_at - -
          Affymetrix Rat Gene 1.0 ST Array 10751352 - -
          Affymetrix Rat Gene 1.0 ST Array 10907524 - -
          Rat Genome U34 Set S76054_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22020445 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1374546 - -
          RatRef-12 Expression BeadChip ILMN_61671 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline