Search
Search
| Gene Symbol: Lima1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191615.2 | NP_001178544.1 | 3 - 4 | 637 | 64 | |
| XM_006257354.2 | XP_006257416.1 | 4 - 5 | 1102 | 64 | ||
| XM_006257355.2 | XP_006257417.1 | 4 - 5 | 785 | 64 | ||
Set Membership: |
|