Search
Search
| Gene Symbol: Bcl9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003749357.3 | XP_003749405.1 | 2 - 3 | 1473 | 68 | |
| XM_006233013.2 | XP_006233075.1 | 2 - 3 | 427 | 68 | ||
| XM_006233014.2 | XP_006233076.1 | 2 - 3 | 427 | 68 | ||
| XM_006233015.2 | XP_006233077.1 | 2 - 3 | 427 | 68 | ||
Set Membership: |
|