Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01533600_m1
          See other CELF3 GE Assays ›
          Gene Symbol:
          Celf3
          Gene Name:
          CUGBP, Elav-like family member 3
          Gene Aliases:
          RGD1563168, Tnrc4
          Chromosome Location:
          Chr.2: 195716360 - 195732872 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01533600_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Celf3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001109190.2 NP_001102660.1 12 - 13 2199 84
          XM_006232897.2 XP_006232959.1 12 - 13 2209 84
          XM_006232898.2 XP_006232960.1 12 - 13 2207 84
          XM_006232900.2 XP_006232962.1 12 - 13 2122 84
          XM_006232902.2 XP_006232964.1 11 - 12 2059 84
          XM_006232903.2 XP_006232965.1 11 - 12 2580 84
          XM_006232904.2 XP_006232966.1 13 - 14 3937 84
          XM_006232905.2 XP_006232967.1 13 - 14 3970 84
          XM_006232906.2 XP_006232968.1 11 - 12 5042 84
          XM_006232907.2 XP_006232969.1 11 - 12 1527 84
          XM_006232908.2 XP_006232970.1 11 - 12 1973 84
          XM_006232911.2 XP_006232973.1 11 - 12 1404 84
          XM_006232912.2 XP_006232974.1 10 - 11 1326 84
          XM_008761322.1 XP_008759544.1 13 - 14 4524 84
          XM_008761323.1 XP_008759545.1 12 - 13 1493 84
          XM_008761324.1 XP_008759546.1 11 - 12 3647 84
          XM_008761325.1 XP_008759547.1 11 - 12 3601 84
          XM_008761326.1 XP_008759548.1 11 - 12 3598 84

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200001684.rn.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of alternative mRNA splicing, via spliceosome
          spermatogenesis
          biological_process
          RNA splicing
          sperm motility
          positive regulation of mRNA splicing, via spliceosome
          nucleotide binding
          molecular_function
          RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P037156 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 640771 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 734801 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 501410 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 537834 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 397419 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 170557 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 267005 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 905841 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 632168 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 96420 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 4503 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P691593 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P691593 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P691593 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 10115 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10817267 - -
          Affymetrix Rat Gene 1.0 ST Array 10817269 - -
          Affymetrix Rat Gene 1.0 ST Array 10817268 - -
          Affymetrix Rat Gene 1.0 ST Array 10817261 - -
          Affymetrix Rat Gene 1.0 ST Array 10817264 - -
          Affymetrix Rat Gene 1.0 ST Array 10817273 - -
          Affymetrix Rat Gene 1.0 ST Array 10817271 - -
          Affymetrix Rat Gene 1.0 ST Array 10817265 - -
          Affymetrix Rat Gene 1.0 ST Array 10817266 - -
          Affymetrix Rat Gene 1.0 ST Array 10817270 - -
          Affymetrix Rat Gene 1.0 ST Array 10817263 - -
          Affymetrix Rat Gene 1.0 ST Array 10817262 - -
          Affymetrix Rat Gene 1.0 ST Array 10817272 - -
          Affymetrix Rat Gene 1.1 ST Array 10817260 - -
          Rat Genome 230 2.0 Array 1382338_PM_at - -
          Rat Genome 230 2.0 Array 1382338_at - -
          Affymetrix HT Rat Focus Array 1382338_PM_at - -
          Affymetrix HT Rat Focus Array 1382338_at - -
          Affymetrix Rat Clariom S Assay PSR0200011249.rn.1 TC0200001684.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200011252.rn.1 TC0200001684.rn.1 RTA TC
          Rat Expression Set 230 1382338_PM_at - -
          Rat Expression Set 230 1382338_at - -
          Affymetrix Rat Gene 1.0 ST Array 10817260 - -
          Rat Genome U34 Set rc_AI102503_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21631201 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1357868 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline