Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01533608_m1
          See other PSMD4 GE Assays ›
          Gene Symbol:
          Psmd4
          Gene Name:
          proteasome (prosome, macropain) 26S subunit, non-ATPase, 4
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 196198303 - 196207568 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01533608_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Psmd4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031331.1 NP_112621.1 2 - 3 239 84
          XM_006232924.2 XP_006232986.1 2 - 3 244 84
          GenBank mRNA AB017188.1 - 2 - 3 239 84
          AF175575.1 - 2 - 3 180 84
          BC060559.1 - 2 - 3 189 6891370 84
          FQ209672.1 - 1 - 2 41 84

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200004195.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Psmd4

          Panther Classification:

          Molecular Function -

          protease

          Gene Ontology Categories:

          Function(s) Process(es)

          ubiquitin-dependent protein catabolic process
          fluid transport
          transcription factor binding
          identical protein binding
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P212708 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 688761 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 193789 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 659097 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 350232 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 75209 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 170839 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 122029 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 160542 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P212708 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P212708 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P212708 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P212708 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700546972 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 45038 - -
          Agilent Rat cDNA Microarray (G4105A) 700546972 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10824918 - -
          Affymetrix Rat Gene 1.0 ST Array 10824919 - -
          Affymetrix Rat Gene 1.0 ST Array 10824920 - -
          Affymetrix Rat Gene 1.0 ST Array 10824915 - -
          Affymetrix Rat Gene 1.0 ST Array 10824924 - -
          Affymetrix Rat Gene 1.0 ST Array 10824923 - -
          Affymetrix Rat Gene 1.0 ST Array 10824925 - -
          Affymetrix Rat Gene 1.0 ST Array 10824922 - -
          Affymetrix Rat Gene 1.0 ST Array 10824921 - -
          Affymetrix Rat Gene 1.0 ST Array 10824916 - -
          Affymetrix Rat Gene 1.1 ST Array 10824914 - -
          Rat Genome 230 2.0 Array 1386930_PM_at - -
          Rat Genome 230 2.0 Array 1386930_at - -
          Affymetrix HT Rat Focus Array 1386930_at - -
          Affymetrix HT Rat Focus Array 1386930_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0200028636.rn.1 TC0200004195.rn.1 RTA TC
          Rat Expression Set 230 1386930_at - -
          Rat Expression Set 230 1386930_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10824914 - -
          Rat Genome U34 Set AB017188_at - -
          Rat Genome U34 Set rc_AA799887_s_at - -
          Rat Genome U34 Set AB017188_g_at - -
          Rat Genome U34 Set rc_AI236731_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21421206 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1353983 - -
          RatRef-12 Expression BeadChip ILMN_49488 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline