Search
Search
| Gene Symbol: Bnipl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128187.2 | NP_001121659.1 | 6 - 7 | 754 | 77 | |
| XM_006232889.2 | XP_006232951.1 | 5 - 6 | 1093 | 77 | ||
| GenBank mRNA | BC166747.1 | - | 6 - 7 | 754 | 5598850 | 77 |
Set Membership: |
|