Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01533928_m1
          See other ITGA10 GE Assays ›
          Gene Symbol:
          Itga10
          Gene Name:
          integrin, alpha 10
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 198772937 - 198792253 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01533928_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Itga10
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001107699.2 NP_001101169.2 27 - 28 3274 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200001772.rn.1

          Set Membership:

          Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          integrin

          Gene Ontology Categories:

          Function(s) Process(es)

          cell-matrix adhesion
          integrin-mediated signaling pathway
          biological_process
          molecular_function
          collagen binding involved in cell-matrix adhesion

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P061012 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P061017 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 256828 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 293548 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 133912 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 959128 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 763237 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 554698 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 471770 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721335 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 952966 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 88946 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 154406 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 781750 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 955396 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 727625 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 824504 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 512738 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 718269 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 40795 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 46495 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 72907 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P061017 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P061012 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P061017 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P061012 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P061012 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P061017 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P215459 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 22998 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10817609 - -
          Affymetrix Rat Gene 1.0 ST Array 10817584 - -
          Affymetrix Rat Gene 1.0 ST Array 10817597 - -
          Affymetrix Rat Gene 1.0 ST Array 10817610 - -
          Affymetrix Rat Gene 1.0 ST Array 10817590 - -
          Affymetrix Rat Gene 1.0 ST Array 10817596 - -
          Affymetrix Rat Gene 1.0 ST Array 10817592 - -
          Affymetrix Rat Gene 1.0 ST Array 10817612 - -
          Affymetrix Rat Gene 1.0 ST Array 10817607 - -
          Affymetrix Rat Gene 1.0 ST Array 10817587 - -
          Affymetrix Rat Gene 1.0 ST Array 10817603 - -
          Affymetrix Rat Gene 1.0 ST Array 10817599 - -
          Affymetrix Rat Gene 1.0 ST Array 10817604 - -
          Affymetrix Rat Gene 1.0 ST Array 10817594 - -
          Affymetrix Rat Gene 1.0 ST Array 10817611 - -
          Affymetrix Rat Gene 1.0 ST Array 10817606 - -
          Affymetrix Rat Gene 1.0 ST Array 10817586 - -
          Affymetrix Rat Gene 1.0 ST Array 10817601 - -
          Affymetrix Rat Gene 1.0 ST Array 10817602 - -
          Affymetrix Rat Gene 1.0 ST Array 10817595 - -
          Affymetrix Rat Gene 1.0 ST Array 10817598 - -
          Affymetrix Rat Gene 1.0 ST Array 10817591 - -
          Affymetrix Rat Gene 1.0 ST Array 10817613 - -
          Affymetrix Rat Gene 1.0 ST Array 10817585 - -
          Affymetrix Rat Gene 1.0 ST Array 10817600 - -
          Affymetrix Rat Gene 1.0 ST Array 10817589 - -
          Affymetrix Rat Gene 1.0 ST Array 10817608 - -
          Affymetrix Rat Gene 1.0 ST Array 10817605 - -
          Affymetrix Rat Gene 1.0 ST Array 10817593 - -
          Affymetrix Rat Gene 1.0 ST Array 10817588 - -
          Affymetrix Rat Gene 1.1 ST Array 10817583 - -
          Affymetrix Rat Clariom S Assay PSR0200012036.rn.1 TC0200001772.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10817583 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22070916 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1373558 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline