Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01534402_m1
          See other GDAP2 GE Assays ›
          Gene Symbol:
          Gdap2
          Gene Name:
          ganglioside-induced differentiation-associated-protein 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 202470469 - 202526849 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01534402_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gdap2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001013201.1 NP_001013219.1 13 - 14 1658 91
          XM_006233032.2 XP_006233094.1 14 - 15 1756 91
          GenBank mRNA BC082000.1 - 13 - 14 1658 7131173 91
          FQ211326.1 - 15 - 16 1839 91

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200001830.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gdap2

          Gene Ontology Categories:

          Function(s)

          response to retinoic acid

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P354053 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 291612 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 483442 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 811022 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 871011 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 778443 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 9044 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 303276 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 256225 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 74604 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 706509 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 769342 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 730890 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 716827 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 32408 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 40922 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 446786 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 282786 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 337230 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 548578 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 192234 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 806213 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 83708 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 684940 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 691088 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 456246 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 437460 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 814712 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 580675 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 478044 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 891162 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 294553 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 464299 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 865772 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 539256 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 395651 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 171863 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 138555 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 174174 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 86391 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 121313 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 26410 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P354053 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P354053 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P354053 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P354053 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 4362 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 23310 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 8018 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 48439 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10817840 - -
          Affymetrix Rat Gene 1.0 ST Array 10817836 - -
          Affymetrix Rat Gene 1.0 ST Array 10817838 - -
          Affymetrix Rat Gene 1.0 ST Array 10817844 - -
          Affymetrix Rat Gene 1.0 ST Array 10817831 - -
          Affymetrix Rat Gene 1.0 ST Array 10817832 - -
          Affymetrix Rat Gene 1.0 ST Array 10817837 - -
          Affymetrix Rat Gene 1.0 ST Array 10817843 - -
          Affymetrix Rat Gene 1.0 ST Array 10817835 - -
          Affymetrix Rat Gene 1.0 ST Array 10817834 - -
          Affymetrix Rat Gene 1.0 ST Array 10817833 - -
          Affymetrix Rat Gene 1.0 ST Array 10817839 - -
          Affymetrix Rat Gene 1.0 ST Array 10817842 - -
          Affymetrix Rat Gene 1.0 ST Array 10817841 - -
          Affymetrix Rat Gene 1.1 ST Array 10817830 - -
          Rat Genome 230 2.0 Array 1379377_PM_at - -
          Rat Genome 230 2.0 Array 1379377_at - -
          Affymetrix HT Rat Focus Array 1379377_at - -
          Affymetrix HT Rat Focus Array 1379377_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0200012521.rn.1 TC0200001830.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200012522.rn.1 TC0200001830.rn.1 RTA TC
          Rat Expression Set 230 1379377_PM_at - -
          Rat Expression Set 230 1379377_at - -
          Affymetrix Rat Gene 1.0 ST Array 10817830 - -
          Rat Genome U34 Set rc_AI178884_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22061774 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1351978 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline