Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01534552_m1
          See other CAB39L GE Assays ›
          Gene Symbol:
          Cab39l
          Gene Name:
          calcium binding protein 39-like
          Gene Aliases:
          -
          Chromosome Location:
          Chr.15: 39745124 - 39849022 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01534552_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cab39l
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001011917.1 NP_001011917.1 9 - 10 1075 87
          XM_006252098.2 XP_006252160.1 10 - 11 1179 87
          XM_006252099.2 XP_006252161.1 11 - 12 1263 87
          XM_006252100.2 XP_006252162.1 12 - 13 1459 87
          XM_006252101.2 XP_006252163.1 11 - 12 1375 87
          XM_008770742.1 XP_008768964.1 11 - 12 1272 87
          XM_008770743.1 XP_008768965.1 8 - 9 1669 87
          GenBank mRNA BC083684.1 - 9 - 10 1075 7188949 87

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1500000587.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cab39l

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P452466 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 897190 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 557144 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 810254 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 488536 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 295724 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 1431 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 475234 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 931562 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 137995 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 323486 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 55744 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 605100 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 786020 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 206849 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 528203 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 772631 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 919583 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 972966 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 593855 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 959090 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 104899 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 696724 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 779550 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 411038 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 464656 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 568047 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 526920 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 401953 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 680414 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 595654 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 68347 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 733769 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 278324 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 366375 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 582139 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 346583 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 722759 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 423154 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 223986 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 375607 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 761840 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 841594 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 289458 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 489545 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 295398 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 727987 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 518353 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 564422 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 42381 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 653222 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 956740 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 415915 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 253438 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 858812 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 957939 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 766463 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 177267 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 180094 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 90920 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 169795 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 148100 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 10668 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 130371 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 138323 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 124834 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P452466 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P452466 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P452466 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P452466 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 104186 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 35731 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 101771 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 24185 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 74435 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10780846 - -
          Affymetrix Rat Gene 1.0 ST Array 10780840 - -
          Affymetrix Rat Gene 1.0 ST Array 10780844 - -
          Affymetrix Rat Gene 1.0 ST Array 10780842 - -
          Affymetrix Rat Gene 1.0 ST Array 10780848 - -
          Affymetrix Rat Gene 1.0 ST Array 10780841 - -
          Affymetrix Rat Gene 1.0 ST Array 10780845 - -
          Affymetrix Rat Gene 1.0 ST Array 10780837 - -
          Affymetrix Rat Gene 1.0 ST Array 10780836 - -
          Affymetrix Rat Gene 1.0 ST Array 10780849 - -
          Affymetrix Rat Gene 1.0 ST Array 10780847 - -
          Affymetrix Rat Gene 1.1 ST Array 10780834 - -
          Rat Genome 230 2.0 Array 1383341_PM_at - -
          Rat Genome 230 2.0 Array 1374094_at - -
          Rat Genome 230 2.0 Array 1374094_PM_at - -
          Rat Genome 230 2.0 Array 1383341_at - -
          Affymetrix HT Rat Focus Array 1383341_PM_at - -
          Affymetrix HT Rat Focus Array 1383341_at - -
          Affymetrix Rat Clariom S Assay PSR1500004147.rn.1 TC1500000587.rn.1 RTA TC
          Rat Expression Set 230 1383341_PM_at - -
          Rat Expression Set 230 1383341_at - -
          Affymetrix Rat Gene 1.0 ST Array 10780834 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21199766 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376640 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1375890 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline