Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01534709_m1
          See other GNRH1 GE Assays ›
          Gene Symbol:
          Gnrh1
          Gene Name:
          gonadotropin-releasing hormone 1 (luteinizing-releasing hormone)
          Gene Aliases:
          Gnrh, Gnrha, Lhrh, Rgnrhg1
          Chromosome Location:
          Chr.15: 44441856 - 44446064 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01534709_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gnrh1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012767.2 NP_036899.1 3 - 4 264 71
          XM_006252096.2 XP_006252158.1 2 - 3 265 71
          GenBank mRNA M12579.1 - 3 - 4 264 71
          S50870.1 - 2 - 3 238 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1500000666.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gnrh1

          Panther Classification:

          Molecular Function -

          peptide hormone intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          multicellular organism development
          female pregnancy
          aging
          regulation of gene expression
          response to organic cyclic compound
          male sex determination
          ovulation
          response to corticosteroid
          response to lipopolysaccharide
          negative regulation of immature T cell proliferation
          response to testosterone
          response to prostaglandin E
          response to potassium ion
          ovulation cycle
          negative regulation of apoptotic process
          response to peptide hormone
          estrous cycle
          response to ethanol
          response to steroid hormone
          response to prolactin
          regulation of ovarian follicle development
          negative regulation of neuron migration
          gonadotropin hormone-releasing hormone activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P206985 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P039752 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 785375 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 481951 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 616370 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 66555 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P206985 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P206985 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P206985 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P529455 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P206985 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 58227 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10781259 - -
          Affymetrix Rat Gene 1.0 ST Array 10784749 - -
          Affymetrix Rat Gene 1.0 ST Array 10781265 - -
          Affymetrix Rat Gene 1.0 ST Array 10784748 - -
          Affymetrix Rat Gene 1.0 ST Array 10781262 - -
          Affymetrix Rat Gene 1.0 ST Array 10784750 - -
          Affymetrix Rat Gene 1.0 ST Array 10781261 - -
          Affymetrix Rat Gene 1.0 ST Array 10781263 - -
          Affymetrix Rat Gene 1.1 ST Array 10781258 - -
          Affymetrix Rat Gene 1.1 ST Array 10784747 - -
          Rat Genome 230 2.0 Array 1370417_PM_at - -
          Rat Genome 230 2.0 Array 1387535_at - -
          Rat Genome 230 2.0 Array 1387535_PM_at - -
          Rat Genome 230 2.0 Array 1370417_at - -
          Affymetrix HT Rat Focus Array 1387535_at - -
          Affymetrix HT Rat Focus Array 1370417_PM_at - -
          Affymetrix HT Rat Focus Array 1387535_PM_at - -
          Affymetrix HT Rat Focus Array 1370417_at - -
          Affymetrix Rat Clariom S Assay PSR1500004691.rn.1 TC1500000666.rn.1 RTA TC
          Rat Expression Set 230 1370417_PM_at - -
          Rat Expression Set 230 1370417_at - -
          Rat Expression Set 230 1387535_at - -
          Rat Expression Set 230 1387535_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10784747 - -
          Affymetrix Rat Gene 1.0 ST Array 10781258 - -
          Rat Genome U34 Set M15527_at - -
          Rat Genome U34 Set M15528mRNA_at - -
          Rat Genome U34 Set M15528mRNA_g_at - -
          Rat Genome U34 Set M12579_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21284867 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1373686 - -
          RatRef-12 Expression BeadChip ILMN_60771 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline