Search
Search
| Gene Symbol: Stmn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270856.1 | NP_001257785.1 | 3 - 4 | 285 | 80 | |
| NM_001270858.1 | NP_001257787.1 | 3 - 4 | 285 | 80 | ||
| GenBank mRNA | AF026528.1 | - | 3 - 4 | 226 | 80 | |
| AF026529.1 | - | 3 - 4 | 226 | 80 | ||
| CB580507.1 | - | 3 - 4 | 216 | 80 | ||
| FQ212976.1 | - | 3 - 4 | 236 | 80 | ||
Set Membership: |
|