Search
Search

Spata13| Gene Symbol: Spata13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191686.1 | NP_001178615.1 | 9 - 10 | 3228 | 108 | |
| XM_006252113.2 | XP_006252175.1 | 9 - 10 | 3295 | 108 | ||
| XM_006252114.2 | XP_006252176.1 | 8 - 9 | 3769 | 108 | ||
| XM_006252115.2 | XP_006252177.1 | 6 - 7 | 2269 | 108 | ||
Set Membership: |
|