Search
Search
| Gene Symbol: Cep44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037181.1 | NP_001032258.1 | 7 - 8 | 1109 | 132 | |
| XM_006253087.2 | XP_006253149.1 | 6 - 7 | 938 | 132 | ||
| XM_006253089.2 | XP_006253151.1 | 7 - 8 | 1083 | 132 | ||
| XM_006253090.2 | XP_006253152.1 | 5 - 6 | 797 | 132 | ||
| XM_008771223.1 | XP_008769445.1 | 7 - 8 | 1368 | 132 | ||
| GenBank mRNA | BC107470.1 | - | 7 - 8 | 1109 | 7457548 | 132 |
| FQ228732.1 | - | 7 - 8 | 1083 | 132 | ||
Set Membership: |
|