Search
Search
| Gene Symbol: Galnt7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022926.1 | NP_075215.1 | 2 - 3 | 634 | 85 | |
| XM_006253083.1 | XP_006253145.1 | 2 - 3 | 659 | 85 | ||
| GenBank mRNA | AF076167.1 | - | 2 - 3 | 634 | 85 | |
Set Membership: |
|