Search
Search
| Gene Symbol: Vipr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017238.1 | NP_058934.1 | 8 - 9 | 922 | 126 | |
| GenBank mRNA | U09631.1 | - | 8 - 9 | 922 | 126 | |
| Z25885.1 | - | 8 - 9 | 875 | 126 | ||
Set Membership: |
|