Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01535822_m1
          See other RBM39 GE Assays ›
          Gene Symbol:
          Rbm39 LOC100910882
          Gene Name:
          RNA binding motif protein 39
          RNA-binding protein 39-like
          Gene Aliases:
          Rnp1, Rnpc2, Rrm
          -
          Chromosome Location:
          Chr.3: 151691505 - 151724715 on Build Rnor_6.0
          Chromosome Location:
          Chr.3: 152226018 - 152259228 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn01535822_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rbm39
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001013207.2 NP_001013225.1 677 85
          XM_006235331.2 XP_006235393.1 735 85
          XM_006235332.2 XP_006235394.1 756 85
          XM_006235335.2 XP_006235397.1 2177 85
          XM_008762349.1 XP_008760571.1 672 85
          XM_008762350.1 XP_008760572.1 693 85
          XM_008762351.1 XP_008760573.1 5 - 6 6575 85
          XM_008762352.1 XP_008760574.1 5 - 6 3667 85
          XM_008762353.1 XP_008760575.1 5309 85
          XM_008762354.1 XP_008760576.1 5 - 6 6596 85
          XM_008762355.1 XP_008760577.1 5085 85
          XM_008762356.1 XP_008760578.1 672 85
          GenBank mRNA BC078917.1 - 750 85
          BC091394.1 - 666 7322533 85
          Gene Symbol: LOC100910882
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_003749605.3 XP_003749653.1 6 - 7 735 85
          XM_003749606.3 XP_003749654.1 6 - 7 735 85
          XM_006235443.2 XP_006235505.1 6 - 7 756 85
          XM_006235446.2 XP_006235508.1 6 - 7 2177 85
          XM_008762415.1 XP_008760637.1 5 - 6 672 85
          XM_008762416.1 XP_008760638.1 5 - 6 693 85
          XM_008762417.1 XP_008760639.1 5 - 6 6575 85
          XM_008762418.1 XP_008760640.1 5 - 6 3667 85
          XM_008762419.1 XP_008760641.1 5 - 6 5309 85
          XM_008762420.1 XP_008760642.1 5 - 6 6596 85
          XM_008762421.1 XP_008760643.1 6 - 7 5085 85
          XM_008762422.1 XP_008760644.1 5 - 6 672 85

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300004569.rn.1, TC0300004575.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rbm39
          Click here to find siRNAs for Gene LOC100910882

          Panther Classification:

          Molecular Function -

          RNA splicing factor

          Gene Ontology Categories:

          Function(s) Process(es)

          mRNA processing
          nucleotide binding
          RNA binding
          poly(A) RNA binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Clariom S Assay PSR0300035297.rn.1 TC0300004575.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300035163.rn.1 TC0300004569.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300035298.rn.1 TC0300004575.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300035164.rn.1 TC0300004569.rn.1 RTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline