Search
Search
| Gene Symbol: Itch | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005887.1 | NP_001005887.1 | 14 - 15 | 1551 | 137 | |
| XM_008762335.1 | XP_008760557.1 | 16 - 17 | 1779 | 137 | ||
| XM_008762336.1 | XP_008760558.1 | 16 - 17 | 1704 | 137 | ||
| XM_008762337.1 | XP_008760559.1 | 13 - 14 | 1456 | 137 | ||
| GenBank mRNA | AY600518.1 | - | 14 - 15 | 1551 | 137 | |
Set Membership: |
|