Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01536253_g1
          See other RGD1307752 GE Assays ›
          Gene Symbol:
          RGD1307752
          Gene Name:
          similar to RIKEN cDNA 1110008F13
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 152933771 - 152943780 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01536253_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: RGD1307752
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001013922.1 NP_001013944.1 3 - 4 498 85
          GenBank mRNA BC089919.1 - 3 - 4 498 7306699 85
          FQ214713.1 - 3 - 4 491 85

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene RGD1307752

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1034346 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 67224 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 787834 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 960694 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 893474 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 810752 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 445806 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 213333 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 127782 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1034346 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1034346 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1034346 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1034346 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 91418 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10841593 - -
          Affymetrix Rat Gene 1.0 ST Array 10841595 - -
          Affymetrix Rat Gene 1.0 ST Array 10873259 - -
          Affymetrix Rat Gene 1.1 ST Array 10841592 - -
          Affymetrix Rat Gene 1.1 ST Array 10873258 - -
          Rat Genome 230 2.0 Array 1371478_PM_at - -
          Rat Genome 230 2.0 Array 1371478_at - -
          Affymetrix HT Rat Focus Array 1371478_PM_at - -
          Affymetrix HT Rat Focus Array 1371478_at - -
          Affymetrix Rat Clariom S Assay PSR0300015996.rn.1 TC0300002162.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300015995.rn.1 TC0300002162.rn.1 RTA TC
          Rat Expression Set 230 1371478_PM_at - -
          Rat Expression Set 230 1371478_at - -
          Affymetrix Rat Gene 1.0 ST Array 10841592 - -
          Rat Genome U34 Set rc_AI011806_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20755991 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1362863 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline